missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitinol tension coil spring, wire diameter 0.75mm (0.030in), mean coil diameter 5.55mm (0.22in), Alloy M
Click to view available options
Quantity:
100 Pc.
25 Pc.
5 Pc.
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nitinol tension coil spring |
| Color | Gray |
| Name Note | Wire diameter 0.75mm, mean coil diameter 5.55mm |
| CAS | 7440-02-0, 7440-32-6 |
| Quantity | 100 Pc. |
| Odor | Odorless |
| Form | Solid |
| Molecular Formula | Ni |
| MDL Number | MFCD00011137 MFCD06798735 |
| Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction