Learn More
Nitinol foil, 0.25mm (0.01in) thick, superelastic, flat annealed, pickled surface
Description
Nitinol foil is used in photo chemically-etched parts, formed sheet shapes, EDM or laser cut shapes, cell-phone technology as a retractable antenna, or microphone boom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Nitinol foil |
| Name Note | 0.25mm (0.01 in.) thick, superelastic, flat annealed, pickled surface |
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Weight | ≈1.02g/25x25mm |
| MDL Number | MFCD02091734 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.