Learn More
Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045492.FI
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Nitinol foil is used in cell-phone technology as a retractable antenna, or microphone boom, due to its highly flexible and mechanical memory nature. It is also used in some novelty products, such as self-bending spoons which can be used by amateur and stage magicians to demonstrate psychic powers or as a practical joke, as the spoon will bend itself when used to stir tea, coffee, or any other warm liquid. It is highly biocompatible and has properties suitable for use in orthopaedic implants. Due to Nitinol's unique properties it has seen a large demand for use in less invasive medical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitinol foil | |
MFCD02091734 | |
Insoluble in water. | |
≈0.25g/25x25mm |
0.05mm (0.002 in.) thick, superelastic, flat annealed, pickled surface | |
50 x 50 mm | |
Odorless |
RUO – Research Use Only