missing translation for 'onlineSavingsMsg'
Learn More

Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.28in), Alloy M, Thermo Scientific™

Product Code. 15494025
Change view
Click to view available options
Quantity:
100 Pc.
25 Pc.
Unit Size:
Pack of 100
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494025 25 Pc. Pack of 25
15404035 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494025 Supplier Thermo Scientific Alfa Aesar Supplier No. 045951.KM
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol compression coil spring, Alloy M
Name Note Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.)
Quantity 25 Pc.
Odor Odorless
MDL Number MFCD02091734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.