missing translation for 'onlineSavingsMsg'
Learn More

NITD008, MedChemExpress

Product Code. 16461319
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16461319

Brand: Medchem Express HY12957

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.

NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.

Specifications

Chemical Name or Material NITD008
CAS 1044589-82-3
Color Gray
Purity Grade Notes Research
Molecular Formula C13H14N4O4
Quantity 1 mg
Synonym 7-Deaza-2'-C-acetylene-adenosine
Solubility Information DMSO : ≥ 50 mg/mL (172.25 mM)
SMILES O[C@@]1(C#C)[C@H](N2C=CC3=C(N)N=CN=C32)O[C@H](CO)[C@H]1O
Molecular Weight (g/mol) 290.27
Formula Weight 290.27
Percent Purity 96.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.