missing translation for 'onlineSavingsMsg'
Learn More

Nital Etchant, 3% (v/v) in Reagent Alcohol, Ricca Chemical

Product Code. 31940487
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940487 1 L Amber Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940487 Supplier Ricca Chemical Company Supplier No. R53162001C
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Chemical Name or Material Nital Etchant, 3% (v/v) in Reagent Alcohol
Color Colorless
CAS 64-17-5, 67-63-0, 67-56-1, 7697-37-2, 7732-18-5
Boiling Point ∼76°C
CAS Min % 85.44, 4.72, 4.27, 3.9, 1.67
CAS Max % 85.44, 4.72, 4.27, 3.9, 1.67
Density 0.81
Odor Alcoholic
Packaging Amber Glass Bottle
pH Acidic
Quantity 1 L
Flash Point 12.5°C
Physical Form Liquid
Solubility Information Miscible in water
Percent Purity 3%
Grade Laboratory
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nital Etchant, 3% (v/v)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
  • P311-Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.