missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16438-1-AP-20UL
This item is not returnable.
View return policy
Description
Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.Specifications
| NIT2 | |
| Polyclonal | |
| Unconjugated | |
| NIT2 | |
| NIT2, nitrilase family, member 2, Nitrilase homolog 2, Omega amidase NIT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288174, 52633, 56954 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q497B0, Q9JHW2, Q9NQR4 | |
| Nit2 | |
| NIT2 Fusion Protein Ag9756 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction