missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595756
This item is not returnable.
View return policy
Description
Immunogen sequence: MLGFITRPPH RFLSLLCPGL RIPQLSVLCA QPRPRAMAIS SSSCELPLVA VCQVTSTPDK QQNFKTCAEL VREAARLGAC LAFLPEAFDF IARDPAETLH LSEPLGGKLL EEYTQLAREC GLWLSLGGFH ERGQDWEQTQ KIYNCHVLLN; Positive Samples: Rat liver; Cellular Location: Cytoplasm, Mitochondrion.
Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.
Specifications
| NIT1 | |
| Polyclonal | |
| Unconjugated | |
| NIT1 | |
| AI255805; Deaminated glutathione amidase; dGSH amidase; ESTM30; MGC57670; Nit1; nitrilase 1; nitrilase homolog 1; rCG_20134; W57327 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 289222 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.27 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TQ94 | |
| NIT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human NIT1 (NP_0011720211). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction