missing translation for 'onlineSavingsMsg'
Learn More

Nisin A, Technical grade (~2.5% mainly sodium chloride and denatured milk solids), TRC

Product Code. 30133729
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133729 25 g 25g
30136028 10 g 10g
30011224 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133729 Supplier Toronto Research Chemicals Supplier No. TRCN48755025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1414-45-5
Synonym 1-Thia-4,7,10,13,16,19-hexaazacyclodocosane, cyclic peptide deriv.∣1-Thia-4,7,10,13-tetraazacyclohexadecane, cyclic peptide deriv.∣1H,9H-Pyrrolo[2,1-i][1,4,7,10]thiatriazacyclotridecine, cyclic peptide deriv.∣9,19-Dithia-2,5,13,16,22-pentaazabicyclo[9.9.2]docosane, cyclic peptide deriv.∣Ambicin N∣Delvoplus∣L-Isoleucyl-(Z)-2,3-didehydro-2-aminobutanoyl-D-cysteinyl-L-isoleucyl-2,3-didehydroalanyl-L-leucyl-L-cysteinyl-threo-3-mercapto-D-2-aminobutanoyl-L-prolylglycyl-L-cysteinyl-L-lysyl-threo-3-mercapto-D-2-aminobutanoylglycyl-L-alanyl-L-leucyl-L-methionylglycyl-L-cysteinyl-L-asparaginyl-L-methionyl-L-lysyl-threo-3-mercapto-D-2-aminobutanoyl-L-alanyl-threo-3-mercapto-D-2-aminobutanoyl-L-cysteinyl-L-histidyl-L-cysteinyl-L-seryl-L-isoleucyl-L-histidyl-L-valyl-2,3-didehydroalanyl-L-lysine Cyclic (3-7),(8-11),(13-19),(23-26),(25-28)-pentakis(sulfide)∣MicroGARD MG 300∣Natural-Keeper∣Nisaplin∣Nisin(1-34)∣Nisitrol∣Novasin

Specifications

Format Neat
Molecular Formula C143H230N42O37S7
Molecular Weight (g/mol) 3354.07, 3354.09
InChI Key NVNLLIYOARQCIX-GSJOZIGCNA-N
SMILES CCC(C)C(N)C(=O)N\C(=C/C)C(=O)NC1CSCC(NC(=O)C(CC(C)C)NC(=O)C(=C)NC(=O)C(NC1=O)C(C)CC)C(=O)NC1C(C)SCC(NC(=O)CNC(=O)C2CCCN2C1=O)C(=O)NC(CCCCN)C(=O)NC1C(C)SCC(NC(=O)CNC(=O)C(CCSC)NC(=O)C(CC(C)C)NC(=O)C(C)NC(=O)CNC1=O)C(=O)NC(CC(N)=O)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC1C(C)SCC2NC(=O)C(NC(=O)C(C)NC1=O)C(C)SCC(NC(=O)C(CC1=CN=CN1)NC2=O)C(=O)NC(CO)C(=O)NC(C(C)CC)C(=O)NC(CC1=CN=CN1)C(=O)NC(C(C)C)C(=O)NC(=C)C(=O)NC(CCCCN)C(O)=O
IUPAC Name 6-amino-2-{2-[2-(2-{2-[2-({7-[6-amino-2-(2-{2-[(21-{6-amino-2-[(10-{15-[(2Z)-2-(2-amino-3-methylpentanamido)but-2-enamido]-12-(butan-2-yl)-9-methylidene-6-(2-methylpropyl)-5,8,11,14-tetraoxo-1-thia-4,7,10,13-tetraazacyclohexadecane-3-amido}-9-methyl-1,4,11-trioxo-tetradecahydropyrrolo[2,1-i]1-thia-4,7,10-triazacyclotridecan-6-yl)formamido]hexanamido}-15,22-dimethyl-12-(2-methylpropyl)-9-[2-(methylsulfanyl)ethyl]-5,8,11,14,17,20-hexaoxo-1-thia-4,7,10,13,16,19-hexaazacyclodocosan-3-yl)formamido]-3-carbamoylpropanamido}-4-(methylsulfanyl)butanamido)hexanamido]-14-[(1H-imidazol-5-yl)methyl]-4,8,20-trimethyl-3,6,12,15,21-pentaoxo-9,19-dithia-2,5,13,16,22-pentaazabicyclo[9.9.2]docosan-17-yl}formamido)-3-hydroxypropanamido]-3-methylpentanamido}-3-(1H-imidazol-5-yl)propanamido)-3-methylbutanamido]prop-2-enamido}hexanoic acid
Quantity 25 g
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.