missing translation for 'onlineSavingsMsg'
Learn More

Nisin, 900 I.U./mg, Thermo Scientific™

Nisin, CAS # 1414-45-5, is a polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria.

Brand:  Thermo Scientific Alfa Aesar J66370.06

Quantity 5g

 View more versions of this product

Product Code. 15497699

  • ONLINE EXCLUSIVE PRICE
    £98.50
    £88.65 / 5g
    Save £9.85
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Nisin is a broad-spectrum polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria
Nisin binds lipid II in the cell membrane. After binding it can insert into the membrane, creating a pore
Nisin can efficiently inhibit the growth of gram-positive bacteria, and resistance is rarely observed
Application
• Nisin is commonly used as a selective compound in cell culture to isolate gram-negative bacteria, yeast, and molds (a type of fungus)
• It has been used in bioassays to study posttranslational modification
• This compound has the ability to bind and interfere with the permeability of the cytoplasmic membrane
• It is used to study the assembly and stability of nisin-lipid II pore complexes
• This compound is used in in vitro experiments to form targeted pores in cell membranes that are mediated by lipid II
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1414-45-5
3354.09
NVNLLIYOARQCIX-OAXMGIFASA-N
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C(
C143H230N42O37S7
MFCD00131724
131847819

Specifications

1414-45-5
5g
C143H230N42O37S7
2967°C
Soluble in water
CCC(C)C1C(=O)NC(=C)C(=O)NC(C(=O)NC(CSCC(C(=O)N1)NC(=O)C(=CC)NC(=O)C(C(C)CC)N)C(=O)NC2C(SCC(NC(=O)CNC(=O)C3CCCN3C2=O)C(=O)NC(CCCCN)C(=O)NC4C(SCC(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC4=O)C)CC(C)C)CCSC)C(=O)NC(CC(=O)N)C(=O)NC(CCSC)C(=O)NC(CCCCN)C(=O)NC5C(
131847819
Nisin, 900 I.U./mg
Brown
>110°C (230°F)
MFCD00131724
14,6564
NVNLLIYOARQCIX-OAXMGIFASA-N
3354.09
3354.06
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.