missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nisin

Product Code. 16443039
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443039 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443039 Supplier MedChemExpress Supplier No. HYP1607
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nisin is a bacteriocin produced by a group of Gram-positive bacteria that belongs to Lactococcus and Streptococcus species.

Specifications

CAS 1414-45-5
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 3354.07 g/mol
Product Type Peptides
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : 1 mg/mL (0.30 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic∣adjust pH to 3 with HCl) (insoluble)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.