missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NISCH Antibody (3B8), Novus Biologicals™

Product Code. 18347119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347119 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347119 Supplier Novus Biologicals™ Supplier No. H00011188M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NISCH Monoclonal antibody specifically detects NISCH in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NISCH
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 3B8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH56900
Gene Alias hIRAS, I-1 receptor candidate protein, I-1FLJ14425, I1R, I1R candidate protein, Imidazoline receptor 1, imidazoline receptor antisera selected, Imidazoline receptor antisera-selected protein, imidazoline receptor candidate, Imidazoline-1 receptor, Imidazoline-1 receptor candidate protein, IR1, IRASFLJ40413, KIAA0975FLJ90519, nischarin
Host Species Mouse
Immunogen NISCH (AAH56900, 1246 a.a. ∽ 1345 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LTGSTPMQVVTCLTRDSYLTHCFLQHLMVVLSSLERTPSPEPVDKDFYSEFGNKTTGKMENYELIHSSRVKFTYPSEEEIGDLTFTVAQKMAEPEKAPAL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11188
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.