missing translation for 'onlineSavingsMsg'
Learn More

Niraparib tosylate, MedChemExpress

Product Code. p-200100293
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485046

Brand: Medchem Express HY10619B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Niraparib tosylate (MK-4827 tosylate) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with an IC50 of 3.8 and 2.1 nM, respectively. Niraparib tosylate leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Niraparib tosylate (MK-4827 tosylate) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with an IC50 of 3.8 and 2.1 nM, respectively. Niraparib tosylate leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Specifications

Chemical Name or Material Niraparib tosylate
CAS 1038915-73-9
Color White
Purity Grade Notes Research
Molecular Formula C26H28N4O4S
Quantity 100 mg
Synonym MK-4827 tosylate
Solubility Information DMSO : 100 mg/mL (203.01 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 1 mg/mL (2.03 mM; heat to 50°C)
SMILES NC(C1=CC=CC2=CN(C3=CC=C([C@H]4CNCCC4)C=C3)N=C21)=O.O=S(C5=CC=C(C)C=C5)(O)=O
Molecular Weight (g/mol) 492.59
Formula Weight 492.59
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.