missing translation for 'onlineSavingsMsg'
Learn More

Niraparib hydrochloride, MedChemExpress

Product Code. 16475046
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475046

Brand: Medchem Express HY10619A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib hydrochloride leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Niraparib hydrochloride (MK-4827 hydrochloride) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib hydrochloride leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity.

Specifications

Chemical Name or Material Niraparib hydrochloride
CAS 1038915-64-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H21ClN4O
Quantity 100 mg
Synonym MK-4827 hydrochloride
Solubility Information DMSO : 250 mg/mL (700.57 mM; Need ultrasonic) ∣H2O : 100 mg/mL (280.23 mM; Need ultrasonic)
SMILES NC(C1=CC=CC2=CN(C3=CC=C([C@H]4CNCCC4)C=C3)N=C21)=O.Cl
Molecular Weight (g/mol) 356.85
Formula Weight 356.85
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.