missing translation for 'onlineSavingsMsg'
Learn More

NIR2 Antibody, Novus Biologicals™

Product Code. 18318948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318948 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318948 Supplier Novus Biologicals Supplier No. H00009600B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

NIR2 Polyclonal antibody specifically detects NIR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. AAH22230.1
Gene Alias DRES9NIR-2, Drosophila retinal degeneration B homolog, FLJ44997, membrane-associated phosphatidylinositol transfer protein 1, NIR2PITPNM, phosphatidylinositol transfer protein, membrane-associated 1Pyk2 N-terminal domain-interacting receptor 2, PITPnm 1, Rd9, RDGB, RDGB1, RDGBA, RDGBA1, retinal degeneration B alpha 1
Host Species Mouse
Immunogen PITPNM1 (AAH22230.1, 1 a.a. - 1243 a.a.) full-length human protein. MLIKEYHILLPMSLDEYQVAQLYMIQKKSREESSGEGSGVEILANRPYTDGPGGSGQYTHKVYHVGSHIPGWFRALLPKAALQVEEESWNAYPYTRTRYTCPFVEKFSIEIETYYLPDGGQQPNVFNLSGAERRQRILDTIDIVRDAVAPGEYKAEEDPRLYHSVKTGRGPLSDDWARTAAQTGPLMCAYKLCKVEFRYWGMQAKIEQFIHDVGLRRVMLRAHRQAWCWQDEWTELSMADIRALEEETARMLAQRMAKCNTGSEGSEAQPPGKPSTEARSAASNTGTPDGPEAPPGPDASPDASFGKQWSSSSRSSYSSQHGGAVSPQSLSEWRMQNIARDSENSSEEEFFDAHEGFSDSEEVFPKEMTKWNSNDFIDAFASPVEAEGTPEPGAEAAKGIEDGAQAPRDSEGLDGAGELGAEACAVHALFLILHSGNILDSGPGDANSKQADVQTLSSAFEAVTRIHFPEALGHVALRLVPCPPICAAAYALVSNLSPYSHDGDSLSRSQDHIPLAALPLLATSSSRYQGAVATVIARTNQAYSAFLRSPEGAGFCGQVALIGDGVGGILGFDALCHSANAGTGSRGSSRRGSMNNELLSPEFGPVRDPLADGVEGLGRGSPEPSALPPQRIPSDMASPEPEGSQNSLQAAPATTSSWEPRRASTAFCPPAASSEAPDGPSSTARLDFKVSGFFLFGSPLGLVLALRKTVMPALEAQMRPACEQIYNLFHAADPCASRLEPLLAPKFQAIAPLTVPRYQKFPLGDGSSLLLADTLQTHSSLFLEELEMLVPSTPTSTSGAFWKGSELATDPPAQPAAPSTTSEVVKILERWWGTKRIDYSLYCPEALTAFPTVTLPHLFHASYWESADVVAFILRQVIEKERPQLAECEEPSIYSPAFPREKWQRKRTQVKIRNVTSNHRASDTVVCEGRPQVLSGRFMYGPLDVVTLTGEKVDVYIMTQPLSGKWIHFGTEVTNSSGRLTFPVPPERALGIGVYPVRMVVRGDHTYAECCLTVVARGTEAVVFSIDGSFTASVSIMGSDPKVRAGAVDVVRHWQDSGYLIVYVTGRPDMQKHRVVAWLSQHNFPHGVVSFCDGLTHDPLRQKAMFLQSLVQEVELNIVAGYGSPKDVAVYAALGLSPSQTYIVGRAVRKLQAQCQFLSDGYVAHLGQLEAGSHSHASSGPPRAALGKSSYGVAAPVDFLRKQSQLLRSRGPSQAEREGPGTPPTTLARGKARSISLKLDSEE
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 9600
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.