missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NIR Dye s775z, NHS
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
NIR Dye s775z, NHS is a near-infrared fluorescent dye; supplied as an NHS ester for coupling to primary amine groups. When conjugated to antibodies, NIR Dye s775z, NHS is substantially brighter and more photostable than competitor dyes. It retains excellent target specificity, even when the degree of labeling (DOL) is very high. Excitation/emission maxima = 775/795 nm; extinction coefficient = 201000 M-1cm-1; quantum yield = 0.09. For further information about how NIR Dye s775z, NHS may be used, please see our protocol: Conjugation Protocol for Amine Reactive Dyes .
Specifications
Specifications
| Chemical Name or Material | 2-((1E,3Z,5E)-4-(2,6-bis((4-(2,5,8,11-tetraoxadodecyl)-1H-1,2,3-triazol-1-yl)methyl)-4-(((2,5-dioxopyrrolidin-1-yl)oxy)carbonyl)phenyl)-7-((E)-3,3-dimethyl-5-sulfonato-1-(3-(trimethylammonio)propyl)indolin-2-ylidene)hepta-1,3,5-trien-1-yl)-3,3-dimethyl-1- |
| Quantity | 1 mg |
| Target | Fluorescent Dyes |
| Molecular Formula | C72H100ClN11O18S2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction