missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP3A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76968
This item is not returnable.
View return policy
Description
NIPSNAP3A Polyclonal specifically detects NIPSNAP3A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| NIPSNAP3A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DKFZp564D177, FLJ13953, HSPC299, MGC14553, nipsnap homolog 3A (C. elegans), NipSnap3A, NipSnap4, protein NipSnap homolog 3A, Protein NipSnap homolog 4, Target for Salmonella secreted protein C, TassC | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25934 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
| NP_056284 | |
| NIPSNAP3A | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human NIPSNAP3A. The immunogen is located within amino acids 40 - 90 of NIPSNAP3A. Amino Acid Squence: EFLENFEKNAHLRTAH | |
| 0.1 mg | |
| Primary | |
| At least two isoforms of NIPSNAP3A are known to exist. NIPSNAP3A antibody is predicted to not cross-react with any other members of the NIPSNAP protein family. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction