missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIPSNAP1 Polyclonal Antibody
Description
A suggested positive control is human brain tissue lysate. PA5-20500 can be used with blocking peptide PEP-0620.
NIPSNAP is a member of an evolutionarily well conserved gene family and has a strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. Recent studies have indicated that NIPSNAP is involved in the regulation of the Ca2+-selective transient receptor potential vanilloid channel 6 (TRPV6). NIPSNAP1 associates with TRPV6 at the plasma membrane and inhibits TRPV6 currents. Other studies show that NIPSNAP's expression is reduced in the phenylketonuria (PKU) mouse brain, suggesting that NIPSNAP may play a role in memory.
Specifications
Specifications
| Antigen | NIPSNAP1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NIPSNAP1 |
| Gene Accession No. | O55125, Q9BPW8 |
| Gene Alias | 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans); nipsnap homolog 1 (C. elegans); Nipsnap1; Protein NipSnap homolog 1 |
| Gene Symbols | NIPSNAP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?