missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPSNAP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76887MFV450
This item is not returnable.
View return policy
Description
NIPSNAP1 Polyclonal antibody specifically detects NIPSNAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NIPSNAP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 450 SE | |
| nipsnap homolog 1 (C. elegans), NIPSNAP, C. elegans, homolog 1,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1, NipSnap1, protein NipSnap homolog 1 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human NIPSNAP. The immunogen is located within the last 50 amino acids of NIPSNAP. Amino Acid Squence: SREETRNAAWRKRGWDEN | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8508 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction