missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£314.00 - £427.00
Specifications
| Antigen | NIPP1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18474332
|
Novus Biologicals
NBP1-90113-25ul |
25 μL |
£314.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256326
|
Novus Biologicals
NBP1-90113 |
0.1 mL |
£427.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NIPP1 Polyclonal specifically detects NIPP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| NIPP1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| activator of RNA decay, ard-1, ARD1nuclear inhibitor of protein phosphatase 1, NIPP1nuclear inhibitor of protein phosphatase-1, NIPP-1nuclear subunit of PP-1, PRO2047, Protein phosphatase 1 regulatory inhibitor subunit 8, protein phosphatase 1 regulatory subunit 8, protein phosphatase 1, regulatory (inhibitor) subunit 8, RNase E | |
| PPP1R8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5511 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ISTLTIEEGNLDIQRPKRKRKNSRVTFSEDDEIINPEDVDPSVGRFRNMVQTAVVPVKKKRVEGPGSLGLEESGSRRMQNFAFSGGLY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title