missing translation for 'onlineSavingsMsg'
Learn More

Nipecotic acid, 98%

Product Code. 11466406
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466406 1 g 1g
11476406 5 g 5g
11486406 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466406 Supplier Thermo Scientific Alfa Aesar Supplier No. B24723.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 498-95-3 | C6H11NO2 | 129.16 g/mol

GABA reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-95-3
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00005992 MFCD01630787
InChI Key XJLSEXAGTJCILF-UHFFFAOYSA-N
Synonym nipecotic acid, 3-piperidinecarboxylic acid, hexahydronicotinic acid, +/--nipecotic acid, h-dl-nip-oh, +/--3-piperidine carboxylic acid, dl-piperidine-3-carboxylic acid, nipecotic, h-nip-oh, 3-carboxypiperidine
PubChem CID 4498
ChEBI CHEBI:116931
IUPAC Name piperidine-3-carboxylic acid
SMILES OC(=O)C1CCCNC1

Specifications

Melting Point ∼261°C (decomposition)
Quantity 1 g
Beilstein 81096
Merck Index 14,6561
Solubility Information Soluble in water
Formula Weight 129.16
Percent Purity 98%
Chemical Name or Material Nipecotic acid
compliance-icons
Product Identifier
  • Nipecotic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Piperidine-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.