missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIPBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18792-1-AP-150UL
This item is not returnable.
View return policy
Description
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.Specifications
| NIPBL | |
| Polyclonal | |
| Unconjugated | |
| NIPBL | |
| CDLS, CDLS1, Delangin, IDN3, IDN3 B, NIPBL, Nipped B homolog (Drosophila), Nipped B like protein, SCC2 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25836, 294787, 71175 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6KC79, Q6KCD5 | |
| Nipbl | |
| NIPBL Fusion Protein Ag5663 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction