missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nipah Virus Glycoprotein G Antibody [PerCP], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43765PCP
This item is not returnable.
View return policy
Description
Nipah Virus Glycoprotein G Polyclonal antibody specifically detects Nipah Virus Glycoprotein G in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Nipah Virus Glycoprotein G | |
| Polyclonal | |
| Western Blot, ELISA | |
| HeV-G | |
| Produced in rabbits immunized with purified, recombinant Nipah Virus Glycoprotein G (RefSeq#: AAY43916.1; Gln71-Thr602) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction