missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564313
This item is not returnable.
View return policy
Description
Immunogen sequence: KEEEIETLNE MSHKLGDPGF VVF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Nipa2 encodes a protein that acts as a selective Mg(2+) transporter. The Nipa2 gene encodes a possible magnesium transporter, and is located adjacent to the imprinted domain in the Prader-Willi syndrome deletion region of chromosome 15. Alternate splicing of the Nipa2 gene results in multiple transcript variants. Pseudogenes of the Nipa2 gene are found on chromosomes 3, 7 and 21. Diseases associated with NIPA2 include Angelman Syndrome and Prader-Willi Syndrome.
Specifications
| NIPA2 | |
| Polyclonal | |
| Unconjugated | |
| NIPA2 | |
| 2600017P10Rik; 3830408P04Rik; AB041581; Magnesium transporter NIPA2; MNCb-2146; Nipa2; non imprinted in Prader-Willi/Angelman syndrome 2; non imprinted in Prader-Willi/Angelman syndrome 2 homolog; non imprinted in Prader-Willi/Angelman syndrome 2 homolog (human); non-imprinted in Prader-Willi/Angelman syndrome region protein 2; Non-imprinted in Prader-Willi/Angelman syndrome region protein 2 homolog; RGD1306051 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N8Q9 | |
| NIPA2 | |
| Recombinant protein corresponding to Human NIPA2. Recombinant protein control fragment (Product #RP-104121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction