missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563783
This item is not returnable.
View return policy
Description
Immunogen sequence: ETRVMFEKIA KYIGENLQLL VDRPDGTYCF RLHNDRVYYV SEKIMKLAAN ISGDKLVSLG TCFGKFTKTH KFRLHVTALD YL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
Specifications
| NIP7 | |
| Polyclonal | |
| Unconjugated | |
| Nip7 | |
| 1110017C15Rik; 60S ribosome subunit biogenesis protein NIP7 homolog; 60S ribosome subunit biogenesis protein NIP-like protein-like protein; 6330509M23Rik; AA408773; AA410017; CGI-37; FLJ10296; HSPC031; HSPC180; hypothetical protein LOC553616; KD93; kDa93; Nip7; NIP7 nucleolar pre-rRNA processing protein; NIP7, nucleolar pre-rRNA processing protein; Nip7p; nuclear import 7; nuclear import 7 homolog; nuclear import 7 homolog (S. cerevisiae); nucleolar pre-rRNA processing protein NIP7; OK/SW-cl.76; OK/SW-cl.78; pEachy; PUA protein; Saccharomyces cerevisiae Nip7p homolog; zgc:110384 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51388 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y221 | |
| Nip7 | |
| Recombinant protein corresponding to Human NIP7. Recombinant protein control fragment (Product #RP-98560). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction