missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NIP30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531986
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, H1299. Predicted reactivity: Mouse (93%), Rat (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FAM192A is also known as Nip30. This protein has not bee characterized.
Specifications
| NIP30 | |
| Polyclonal | |
| Unconjugated | |
| FAM192A | |
| 1700001O11Rik; 2310065K24Rik; AI596259; C16orf94; CDA018; CDA10; Fam192a; family with sequence similarity 192 member A; family with sequence similarity 192, member A; NEFA-interacting nuclear protein NIP30; Nip30; PA28G-interacting protein; PIP30; protein FAM192A; PSME3-interacting protein; RGD1307433 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.42 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9GZU8 | |
| FAM192A | |
| Recombinant protein encompassing a sequence within the center region of human NIP30. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction