missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | NIP |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18155129
|
Novus Biologicals
NBP2-47356 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649385
|
Novus Biologicals
NBP2-47356-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NIP Polyclonal specifically detects NIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NIP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 90527 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VAHRMQPHRLKAFFNQSVDEDPMLEWSPEEGGLLSPRYRSMADSPKSQDIPLSEASS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Dual oxidase activator 1, dual oxidase maturation factor 1, dual oxidase maturation factor 1 delta, dual oxidase maturation factor 1 gamma, FLJ32334, homolog of Drosophila Numb-interacting protein, mol, NIPdual oxidase maturation factor 1 alpha, Numb-interacting protein, NUMBIPdual oxidase maturation factor 1 beta | |
| DUOXA1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title