missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxide, Puratronic™, 99.9985% (metals basis)

Product Code. 11396947
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396947 250 g 250g
11306957 50 g 50g
11316957 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11396947 Supplier Thermo Scientific Alfa Aesar Supplier No. 010822.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1313-96-8 | Nb2O5 | 265.81 g/mol

Niobium pentoxide is the precursor to a majority of niobium compounds, powder metallurgy products, and alloys. It is also used in glass, optic and ceramic applications. The nanoparticles of niobium pentoxide are useful in the preparation of electrochemical capacitors and cathodes. In addition, appropriate preparations of this compound can be used as a dietary supplement in food industry. It is a precursor to lithium niobate which finds extensive use in piezoelectric sensors, anti-aliasing filters, optical waveguides, mobile phones, surface acoustic devices, optical modulators, including linear and non-linear optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-96-8
Molecular Formula Nb2O5
Molecular Weight (g/mol) 265.81
MDL Number MFCD00011128
InChI Key ZKATWMILCYLAPD-UHFFFAOYSA-N
Synonym niobium oxide, niobium v oxide, niobium pentoxide, diniobium pentaoxide, niobia, niobium pentaoxide, diniobium pentoxide, niobium 5+ oxide, nobum oxde
PubChem CID 123105
SMILES O=[Nb](=O)O[Nb](=O)=O

Specifications

Melting Point 1520°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9985% (metals basis)
Solubility Information Insoluble in water; soluble in hydrogen fluoride.
IUPAC Name dioxoniobiooxy(dioxo)niobium
Formula Weight 265.81
Odor Odorless
Appearance White powder or lump
Density 4.6
Chemical Name or Material Niobium(V) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.