missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxalate hydrate

Product Code. p-4412876
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11410121

Brand: Thermo Scientific Alfa Aesar 044819.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1660137-09-6 | C10H12NbO21 | 561.091 g/mol

Niobium(V) oxalate hydrate is used as doping precursors in the PZT composition to improve its piezoelectric properties. It is also used in the preparation of niobium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660137-09-6
MDL Number MFCD00050743
Synonym niobium v oxalate, niobium oxalate
PubChem CID 121233648

Specifications

Physical Form Powder
Quantity 25 g
Molecular Formula C10H12NbO21
InChI Key CRIYBCSTIVPHBQ-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Nb]
IUPAC Name niobium;oxalic acid;hydrate
Molecular Weight (g/mol) 561.091
Formula Weight 538.04 (Anhydrous)
Odor Odorless
Chemical Name or Material Niobium(V) oxalate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.