missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) oxalate hydrate

Product Code. 11420121
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420121 100 g 100g
11410121 25 g 25g
11430121 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420121 Supplier Thermo Scientific Alfa Aesar Supplier No. 044819.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1660137-09-6 | C10H12NbO21 | 561.091 g/mol

Niobium(V) oxalate hydrate is used as doping precursors in the PZT composition to improve its piezoelectric properties. It is also used in the preparation of niobium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660137-09-6
MDL Number MFCD00050743
Synonym niobium v oxalate, niobium oxalate
PubChem CID 121233648

Specifications

Physical Form Powder
Quantity 100 g
Molecular Formula C10H12NbO21
InChI Key CRIYBCSTIVPHBQ-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Nb]
IUPAC Name niobium;oxalic acid;hydrate
Molecular Weight (g/mol) 561.091
Formula Weight 538.04 (Anhydrous)
Odor Odorless
Chemical Name or Material Niobium(V) oxalate hydrate
compliance-icons
Product Identifier
  • Niobium(V) oxalate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Niobium(V) oxalate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.