missing translation for 'onlineSavingsMsg'
Learn More

Niobium(V) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

£78.20 - £510.00

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.156
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011548.14
25 g
£78.20
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11327257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011548.22
100 g
£184.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11317257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011548.36
500 g
£510.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10026-12-7
270.156
YHBDIEWMOMLKOO-UHFFFAOYSA-I
24818
Cl[Nb](Cl)(Cl)(Cl)Cl
Cl5Nb
MFCD00011127
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
pentachloroniobium

Specifications

248.2°C
10026-12-7
Powder
Cl5Nb
niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
YHBDIEWMOMLKOO-UHFFFAOYSA-I
pentachloroniobium
24818
Acidic
Yellow powder/flake/crystalline/beads
Niobium(V) chloride
208.3°C
White
99.9% (metals basis)
MFCD00011127
Soluble in alcohol,hydrochloric acid,carbon tetrachloride and chloroform.
Cl[Nb](Cl)(Cl)(Cl)Cl
270.156
270.2
Moisture sensitive
2.74
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 233-059-8

missing translation for 'rtecsNumber' : QU0350000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only