missing translation for 'onlineSavingsMsg'
Learn More

Niobium solution for ICP 1g/L HNO3/HFmatrix, Fisher chemical™
BIOPHARMA

Product Code. 12953454 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12953454 100 mL Bottle 100mL
12963454 250 mL Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 12953454 Supplier Fisher Chemical Supplier No. S 036 100
  • ONLINE EXCLUSIVE PRICE
    £57.05
    £56.04 / 100mL
    Save £1.01
    1% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Nb
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Nb
Solvent or Matrix Nitric Acid/Hydrogen Fluoride
Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Niobium (Nb) 1000mg/l in HNO3 5%,HF 1%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.