missing translation for 'onlineSavingsMsg'
Learn More

Niobium slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number 11380068
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Percent Purity 99.95%
Odor Odorless
Weight ≈1.7g/slug
Form Slug
Assay (metals basis excluding Ta)
Chemical Name or Material Niobium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'