missing translation for 'onlineSavingsMsg'
Learn More

Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 11328458
Click to view available options
Quantity:
10 g
100 g
250 g
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]
This item is not returnable. View return policy
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium, element, niobium-93, powder, foil, rod, vn 1, foil, 0.25mm 0.01in thick, columbio, niobio
23936
CHEBI:33344
niobium
[Nb]

Used in high temperature and super alloys of Zn and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance gray powder
Density 8.57g/cm3 at 20°C
Assay Percent Range 99.8% (Metals basis)
Quantity 10 g
Odor Odorless
Form Powder
Chemical Name or Material Niobium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'