missing translation for 'onlineSavingsMsg'
Learn More

Niobium phenoxide, Thermo Scientific™

Product Code. p-7080280
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
Product Code. Quantity unitSize
11438650 0.5 g 0.5g
11428650 2 g 2g
11418650 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 11438650

Brand: Thermo Scientific Alfa Aesar 051120.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16576-63-9
Molecular Formula C30H25NbO5
Molecular Weight (g/mol) 558.431
MDL Number MFCD00014076
InChI Key ZCNDFHXTXLMYDM-UHFFFAOYSA-I
Synonym niobium 5+ pentakisphenoxide, pentaphenoxyniobium v, niobium 5+ ion pentaphenolate, columbium penta phenoxide, niobium 5+ pentaphenolate, phenol, niobium 5+ salt 5:1
PubChem CID 25021451
IUPAC Name niobium(5+);pentaphenoxide
SMILES C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].C1=CC=C(C=C1)[O-].[Nb+5]

Specifications

Melting Point 208°C
Linear Formula Nb(C6H5O)5
Quantity 0.5 g
Sensitivity Moisture sensitive
Formula Weight 558.44
Physical Form Powder
Chemical Name or Material Niobium phenoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.