missing translation for 'onlineSavingsMsg'
Learn More

Niobium nitride

Product Code. p-7048936
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11336518

Brand: Thermo Scientific Alfa Aesar 012146.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24621-21-4 | NNb | 106.913 g/mol

Niobium nitride is used as superconductor. Used in absorbing anti-reflective coatings. It used to produce arc-welding rods and in steel. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24621-21-4
Molecular Formula NNb
Molecular Weight (g/mol) 106.913
MDL Number MFCD00016239
InChI Key CFJRGWXELQQLSA-UHFFFAOYSA-N
Synonym niobium nitride, niobium nitride nbn, columbium nitride, niobiumylidyneamine, niobium nitride 10g
PubChem CID 90560
IUPAC Name azanylidyneniobium
SMILES N#[Nb]

Specifications

Melting Point 2573°C
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 106.92
Odor Odorless
Density 8.40 g/mL
Chemical Name or Material Niobium nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.