missing translation for 'onlineSavingsMsg'
Learn More

Niobium n-butoxide, 99% (metals basis)

Product Code. 11377998
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377998 25 g 25g
11357998 1 g 1g
11367998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377998 Supplier Thermo Scientific Alfa Aesar Supplier No. 036570.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51030-47-8 | C20H50NbO5 | 463.521 g/mol

Niobium n-butoxide plays an important role as a precursor in chemical synthesis. It is also used in the production of nano-materials. It acts as a precursor for dielectric films and structures with controlled properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51030-47-8
Molecular Formula C20H50NbO5
Molecular Weight (g/mol) 463.521
MDL Number MFCD00144916
InChI Key OMBQVKDAXPUDBD-UHFFFAOYSA-N
Synonym butan-1-ol; niobium, niobium v-n-butoxide
PubChem CID 50912030
IUPAC Name butan-1-ol;niobium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.CCCCO.[Nb]

Specifications

Density 1.09 g/mL
Boiling Point 197°C (0.15 mmHg)
Linear Formula Nb[O(CH2)3CH3]5
Quantity 25 g
Sensitivity Moisture sensitive
Solubility Information Miscible with hydrocarbons,ketones and esters.
Formula Weight 458.12
Percent Purity 99%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium n-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.