missing translation for 'onlineSavingsMsg'
Learn More

Niobium(IV) oxide, 99+% (metals basis)

Product Code. 11493810
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493810 10 g 10g
11483810 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493810 Supplier Thermo Scientific Alfa Aesar Supplier No. 089692.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12034-59-2 | NbO2 | 124.90 g/mol

Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12034-59-2
Molecular Formula NbO2
Molecular Weight (g/mol) 124.90
MDL Number MFCD00016242
InChI Key HFLAMWCKUFHSAZ-UHFFFAOYSA-N
Synonym niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
PubChem CID 82839
IUPAC Name dioxoniobium
SMILES O=[Nb]=O

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range >99% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 124.91
Density 5.9 g/mL
Chemical Name or Material Niobium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.