missing translation for 'onlineSavingsMsg'
Learn More

Niobium isopropoxide, 99% (metals basis), 10% w/v in isopropanol/hexane (50:50)

Product Code. 11308008
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308008 25 mL 25mL
11318008 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308008 Supplier Thermo Scientific Alfa Aesar Supplier No. 036572.AC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18368-80-4 | C15H40NbO5 | 393.386 g/mol

Niobium isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-80-4
Molecular Formula C15H40NbO5
Molecular Weight (g/mol) 393.386
MDL Number MFCD00144915
InChI Key FTYCGXPVPAOYKN-UHFFFAOYSA-N
Synonym niobium isopropoxide, in isopropanol/hexane
PubChem CID 76420078
IUPAC Name niobium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Nb]

Specifications

Linear Formula Nb[OCH(CH3)2]5
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information It reacts with water. Soluble in hydrocarbons and warm isopropanol,reacts with other alcohols,ketones and esters.
Formula Weight 388.35
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 10% w/v in isopropanol/hexane (50:50)
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.