missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nintedanib, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
Description
Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Nintedanib |
| CAS | 656247-17-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C31H33N5O4 |
| Quantity | 200 mg |
| Synonym | BIBF 1120 |
| Solubility Information | DMSO : 11.5 mg/mL (21.31 mM; Need ultrasonic) |
| SMILES | O=C1NC2=CC(C(OC)=O)=CC=C2/C1=C(NC3=CC=C(N(C(CN4CCN(C)CC4)=O)C)C=C3)\C5=CC=CC=C5 |
| Molecular Weight (g/mol) | 539.62 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction