Learn More
Invitrogen™ NINL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596726
Description
Immunogen sequence: MDEEENHYVS QLREVYSSCD TTGTGFLDRQ ELTQLCLKLH LEQQLPVLLQ TLLGNDHFAR VNFEEFKEGF VAVLSSNAGV RPSDEDSSSL ESAASSAIPP KYVNGSKWYG RRSRPELCDA ATEARRVPEQ QTQASLKSHL WRSASLESVE SPKSDEEAES TKEAQNELFE AQGQLQTWDS EDFGSPQKSC; Positive Samples: Mouse testis; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center.
Involved in the microtubule organization in interphase cells. Overexpression induces the fragmentation of the Golgi, and causes lysosomes to disperse toward the cell periphery; it also interferes with mitotic spindle assembly. May play a role in ovarian carcinogenesis.
Specifications
| NINL | |
| Polyclonal | |
| Unconjugated | |
| NINL | |
| 4930519N13Rik; Gm1004; Gm1634; Kiaa0980; LOW QUALITY PROTEIN: ninein-like protein; mKIAA0980; ninein like; ninein-like; ninein-like protein; NINL; Nlp; RGD1306152 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 78177 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZQ12 | |
| NINL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human NINL (NP_0794523). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.