missing translation for 'onlineSavingsMsg'
Learn More

Skytec Ninja Silver+ Gloves

Product Code. 17832164
Click to view available options
Size:
6, X-Small
7, Small
8, Medium
9, Large
10, X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ninja Silver+ Gloves offer excellent cut resistance whilst maintaining a high degree of comfort and dexterity. PU coating protects the hand from oils and grease delivering effective long lasting grip.

  • Level C cut resistance (EN388:2016)
  • Excellent grip in wet, dry or sligtly oily conditions
  • Extremely durable with high tear and abrasion resistance
  • Retains flexibility despite its high protection levels
  • Breathable liner
  • Elasticated wrist provides a secure and comfortable fit
  • Mottled grey colouring extends wearable life

Specifications

Certifications/Compliance CE CAT II, ANSI Cut
Certifications/Compliance (Europe) EN388, EN420
Coated Area Palm
Construction Machine Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material HPPE, Nylon, Glass Fiber, Polyurethane
Product Type Knitted Dipped Glove
Size 10, X-Large
EN388 Standard 4X43C
For Use With (Application) Cut Protection
Gender Unisex
Glove Cuff Style Elasticated
Liner Material HPPE, Nylon, Glass Fiber
Lining Lined
Manufacturing/QMS Audit Standards ISO9001:2015
Packaging Type Bag
Series Ninja Silver+
Thumb Style Straight
Yarn Gauge 13 G
Length (English) 9 in.
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'