missing translation for 'onlineSavingsMsg'
Learn More

Skytec Ninja Flex Work Gloves

Product Code. 17862194
Change view
Click to view available options
Size:
7, Small
8, Medium
9, Large
10, X-Large
Unit Size:
10 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17862194 7, Small 10 pairs
17852194 8, Medium 10 pairs
17842164 9, Large 10 pairs
17892194 10, X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17862194 Supplier Skytec Supplier No. SKY191
Outlet Corner
Use promo code 28080
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Ninja Flex Work Gloves feature seamless lightweight liner and latex coating that maintain an overall superthin feel for enhanced dexterity, fit and comfort. For manual handling in damp and dry conditions.

  • The lightweight nylon/lycra liner (15 gauge) maintains a superthin 'feel' for improved dexterity, fit and comfort
  • Anatomically designed for high levels of comfort and tactile touch
  • Abrasion and rip resistant
  • Excellent grip in both wet and dry environments
  • Silicone and lint free
  • Seamless liner prevents irritation

Specifications

Certifications/Compliance CE CAT II
Certifications/Compliance (Europe) EN388, EN420
Chlorinated No
Coated Area Palm
Construction Knit
Coating Material Natural Latex
Color Red, Gray
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Nylon, Latex
Product Type Grip Glove
Size 7, Small
For Use With (Application) General handling
Glove Cuff Style Elasticated
Resistant To Abrasion
Thumb Style Straight
Length (English) 9 in.
Length (Metric) 240 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.