missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ninhydrin, spectrophotometric grade

Product Code. 11964861 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11964861

Brand: Thermo Scientific Chemicals 165871000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym 1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Melting Point 250.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in ether and chloroform
Infrared Spectrum Authentic
Formula Weight 178.14
Grade Spectrophotometric
Packaging Glass bottle
Suitability (for amino acid analysis): passes
Density 0.81
Color Yellow
pH 4.6 to 5.6 (1% aq. soln.)
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ninhydrin
Show More Show Less
compliance-icons
Product Identifier
  • Ninhydrin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.