missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ninhydrin, ozone friendly ready to use spray for TLC

Product Code. 10780971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
240 mL
Unit Size:
240mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10780971 240 mL 240mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10780971 Supplier Thermo Scientific Chemicals Supplier No. 327022400
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ninhydrin
Boiling Point >34.0°C
CAS 485-47-2, 106-97-8, 74-98-6, 71-36-3
Packaging Spray can
Molecular Formula C9H6O4
MDL Number MFCD00003791
Density 0.81
Merck Index 15, 6640
Quantity 240 mL
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.143
PubChem CID 10236
ChEBI CHEBI:86374
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ninhydrin, ozone friendly ready to use spray for TLC
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P251-Do not pierce or burn, even after use.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: n-Butyl alcohol, L.P.G. (liquified petroleum gas), 1H-Indene-1, 3(2H)-dione, 2,2-dihydroxy-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.