Learn More
Thermo Scientific Chemicals Ninhydrin, ACS reagent
Description
Reagent for determination of amino acids and amine containing compounds, as an intermediate.Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Reagent for determination of amino acids and amine containing compounds, as an intermediate
Specifications
Specifications
| Melting Point | 251°C (decomposition) |
| Beilstein | 1910963 |
| Merck Index | 14,6554 |
| Solubility Information | Soluble in water,acetone,ethanol,and methanol; Slightly soluble in chloroform and ether. |
| Formula Weight | 178.14 |
| Grade | ACS |
| Sensitivity | Light sensitive |
| Color | Brown to Yellow |
| Quantity | 100 g |
| Physical Form | Crystalline |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.