missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74631
This item is not returnable.
View return policy
Description
Ninein Polyclonal specifically detects Ninein in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Ninein | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein, hNinein, KIAA1565, ninein, ninein (GSK3B interacting protein), ninein centrosomal protein | |
| Rabbit | |
| 243 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q8N4C6 | |
| NIN | |
| The immunogen recognized by this antibody maps to a region between residue 1850 and 1900 of human Ninein using the numbering given in entry NP_891991.1 (GeneID 51199). | |
| Affinity Purified | |
| RUO | |
| 51199 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction