missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93990-0.02ml
This item is not returnable.
View return policy
Description
Ninein Polyclonal antibody specifically detects Ninein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Ninein | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein, hNinein, KIAA1565, ninein, ninein (GSK3B interacting protein), ninein centrosomal protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human NIN (NP_891991.1). MDEVEQDQHEARLKELFDSFDTTGTGSLGQEELTDLCHMLSLEEVAPVLQQTLLQDNLLGRVHFDQFKEALILILSRTLSNEEHFQEPDCSLEAQPKYVRGGKRYGRRSLPEFQESVEEFPEVTVIEPLDEEARPSHIPAGDCSEHWKTQRSEEYEAEGQLRFWNPDDLNASQSGSSPPQDWIEEKLQEVCEDLGITRDGHLNRKKLVSICEQYGLQNVDGEMLEEVFHNLDPDGTMSVEDFFYGLFKNGKSLTPSASTPYRQLKRHLSMQSFDESGRRTTTSSAMT | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 51199 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction