missing translation for 'onlineSavingsMsg'
Learn More

Nimodipine, 98+%

Product Code. 15493899
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493899 1 g 1g
15413909 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493899 Supplier Thermo Scientific Alfa Aesar Supplier No. J61287.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent L-type calcium channel antagonist | CAS: 66085-59-4 | C21H26N2O7 | 418.45 g/mol

Potent L-type calcium channel antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66085-59-4
Molecular Formula C21H26N2O7
Molecular Weight (g/mol) 418.45
MDL Number MFCD00153848
InChI Key UIAGMCDKSXEBJQ-UHFFFAOYNA-N
Synonym nimodipine, nimotop, periplum, nimodipino, nimodipinum, nymalize, admon, nimodipinum inn-latin, nimodipino inn-spanish, isopropyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate
PubChem CID 4497
ChEBI CHEBI:7575
IUPAC Name 3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C

Specifications

Color Yellow
Quantity 1 g
Merck Index 14,6551
Formula Weight 418.44
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Nimodipine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.