missing translation for 'onlineSavingsMsg'
Learn More

Nimesulide, MedChemExpress

Product Code. 16478256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478256 100 mg 100mg
16442116 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478256 Supplier Medchem Express Supplier No. HYB0363
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nimesulide is a selective COX-2 inhibitor, with IC50s of 70 nM-70 μM in a time-dependent manner, but it shows no effect on COX-1 (IC50 >100 μM). Nimesulide has potent anti-inflammatory, analgesic and antipyretic properties.

Nimesulide is a selective COX-2 inhibitor, with IC50s of 70 nM-70 μM in a time-dependent manner, but it shows no effect on COX-1 (IC50 >100 μM). Nimesulide has potent anti-inflammatory, analgesic and antipyretic properties.

Specifications

Chemical Name or Material Nimesulide
CAS 51803-78-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C13H12N2O5S
Quantity 100 mg
Synonym R805
Solubility Information DMSO : ≥ 100 mg/mL (324.35 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CS(=O)(NC1=CC=C([N+]([O-])=O)C=C1OC2=CC=CC=C2)=O
Molecular Weight (g/mol) 308.31
Formula Weight 308.31
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.